Sign In to View Organizational & Contract Pricing.
Select a Size
Change View
| Size/SKU | Availability | Price |
|---|
About This Item
Empirical Formula (Hill Notation):
C18H14N4O3S
CAS Number:
Molecular Weight:
366.39
UNSPSC Code:
12352200
Assay:
≥98% (HPLC)
Form:
powder
Pricing and availability is not currently available.
Quality Segment
assay
≥98% (HPLC)
form
powder
color
white to beige
solubility
DMSO: 2 mg/mL, clear (warmed)
storage temp.
2-8°C
Biochem/physiol Actions
SPC-839 is a potent suppressors of T cell activation that covalently targets XPO1 at Cys 528. It potently inhibits inflammation in mice with minimal impact on nuclear export and cell viability. SPC-839 exhibits efficacy in in vivo models of autoimmune disease.
Storage Class
11 - Combustible Solids
flash_point_f
Not applicable
flash_point_c
Not applicable
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.