Select a Size
| Size/SKU | Availability | Price |
|---|
About This Item
grade
USP Analytical Reference Materials (ARMs)
agency
USP
form
powder
packaging
pkg of 200 mg
manufacturer/tradename
USP
application(s)
pharmaceutical small molecule
SMILES string
O=C1C=C(NCCCN2CCN(C=3C=CC=CC3OC)CC2)N(C(=O)N1C)C
InChI
InChI=1S/C20H29N5O3/c1-22-18(15-19(26)23(2)20(22)27)21-9-6-10-24-11-13-25(14-12-24)16-7-4-5-8-17(16)28-3/h4-5,7-8,15,21H,6,9-14H2,1-3H3
InChI key
ICMGLRUYEQNHPF-UHFFFAOYSA-N
General description
Analysis Note
1 of 1
This Item | |||
|---|---|---|---|
| description United States Pharmacopeia (USP) Reference Standard | description - | description European Pharmacopoeia (EP) Reference Standard | description - |
| application(s) pharmaceutical small molecule | application(s) - | application(s) pharmaceutical | application(s) - |
| form powder | form - | form - | form - |
| grade USP Analytical Reference Materials (ARMs) | grade - | grade pharmaceutical primary standard | grade - |
| manufacturer/tradename USP | manufacturer/tradename - | manufacturer/tradename EDQM | manufacturer/tradename - |
| packaging pkg of 200 mg | packaging - | packaging - | packaging - |
| agency USP | agency - | agency - | agency - |
Signal Word
Warning
Hazard Codes
Precautionary Statements
Hazard Classifications
Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3
Target Organs
Respiratory system
Storage Class
11 - Combustible Solids
Flash Point (°F)
Not applicable
Flash Point (°C)
Not applicable
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.




