Merck
  • Identification of the human liver microsomal cytochrome P450s involved in the metabolism of N-nitrosodi-n-propylamine.
Identification of the human liver microsomal cytochrome P450s involved in the metabolism of N-nitrosodi-n-propylamine.
Carcinogenesis (2000-07-27)
J F Teiber, P F Hollenberg
ABSTRACT
The ability of human liver cytochrome P450s to metabolize the environmental carcinogen N-nitrosodi-n-propylamine (NDPA) was investigated. The maximum rate of NDPA depropylation in seven human liver microsomal samples was 1.15 nmol/min/mg (range 0.53-2.60). Troleandomycin, a P450 3A4/5 inhibitor, inhibited depropylation modestly (10-60%) in three of seven samples. Diethyldithiocarbamic acid, a potent 2E1 inhibitor, and a 2E1 inhibitory monoclonal antibody (mAb) inhibited the reaction in all samples (23 to almost 100%). No significant inhibition was observed with the 2C9 inhibitor sulfaphenazole or with mAbs to 3A4, 2A6 and 2D6. The 2C8/9/18/19 mAb inhibited depropylation in one sample by approximately 25% and approximately 25% of the activity in another sample could not be accounted for by the inhibitors. Denitrosation of NDPA by three of the microsomal samples exhibited low K(m) values (51-86 microM) while two of these also had high K(m) values (2.6 and 4.6 mM). Purified human P450 2B6 and 3A4 and human P450 2A6, 2C8, 2C9 and 2D6 membranes had high K(m) values relative to their maximum turnover rates and are unlikely to participate in NDPA metabolism at micromolar concentrations. Conversely, purified rabbit 2E1 exhibited K(m) and V(max) values for depropylation of 52 microM and 13.4 nmol propionaldehyde/min/nmol P450, respectively. Values for denitrosation were 66 microM and 1.44 nmol nitrite/min/nmol P450, respectively. The toxicity of NDPA in transfected human liver epithelial cells expressing 2E1 was dose dependent down to 50 microM. No toxicity was observed in control cells or those expressing 2A6. These results indicate that 2E1 is the major human liver microsomal isoform responsible for NDPA metabolism at low micromolar concentrations. We also show that purified P450s catalyze the denitrosation of NDPA at approximately 10-20% of the rate of depropylation and K(m) values for both reactions are the same for each isozyme. This is consistent with the formation of an initial intermediate common to both pathways, presumably an alpha-nitrosamino radical.
Product Description
Supelco
N-Nitrosodi-n-propylamine solution, certified reference material, 5000 μg/mL in methanol
Supelco
N-Nitrosodi-n-propylamine, analytical standard