Merck
Development of MRI phantom equivalent to human tissues for 3.0-T MRI.
Medical physics (2013-03-08)
Kengo Hattori, Yusuke Ikemoto, Wataru Takao, Seiichiro Ohno, Takashi Harimoto, Susumu Kanazawa, Masataka Oita, Koichi Shibuya, Masahiro Kuroda, Hirokazu Kato
ABSTRACT
A 3.0-T MRI phantom (called the CAGN-3.0T phantom) having human-equivalent relaxation times and human-equivalent conductivity was developed. The ingredients of the phantom are carrageenan (as a gelatinizer), agarose (as a T2-relaxation modifier), GdCl3 (as a T1-relaxation modifier), NaCl (as a conductivity modifier), and NaN3 (as an antiseptic). Numerous samples with varying concentrations of agarose, GdCl3, and NaCl were prepared, and T1 and T2 values were measured using 3.0-T MRI. The T1 values of the CAGN-3.0T phantom were unaffected by NaCl, while the T2 values were only slightly affected. Based on the measured data, empirical formulae were devised to express the relationships between the concentrations of agarose, GdCl3, and NaCl and the relaxation times. The formula for expressing the conductivity of the CAGN-3.0T phantom was obtained. By adjustments to the concentrations of agarose, GdCl3, and NaCl, the relaxation times and conductivity of almost all types of human tissues can be simulated by CAGN-3.0T phantoms. The phantoms have T1 values of 395-2601 ms, T2 values of 29-334 ms, and conductivity of 0.27-1.26 S/m when concentrations of agarose, GdCl3, and NaCl are varied from 0 to 2.0 w/w%, 0 to 180 μmol/kg, and 0 to 0.7 w/w%, respectively. The CAGN-3.0T phantom has sufficient strength to replicate the torso without using reinforcing agents, and can be cut with a knife into any shape.
Sigma-Aldrich
Sepharose 4B, 45-165 μm bead diameter
Sigma-Aldrich
Gadolinium(III) chloride, powder, 99.99% trace metals basis
Sigma-Aldrich
Agarose, Type I, low EEO
Sigma-Aldrich
Agarose, Medium EEO, Molecular Biology
Sigma-Aldrich
Agarose, Ultra-low Gelling Temperature, molecular biology grade
Sigma-Aldrich
Agarose, Low EEO
Sigma-Aldrich
Agarose, High EEO
Sigma-Aldrich
Agarose, Low EEO, for Immunoelectrophoresis
Sigma-Aldrich
Agarose, Type II-A, Medium EEO
Sigma-Aldrich
Agarose, Wide range, Molecular Biology
Sigma-Aldrich
Agarose, High EEO
Sigma-Aldrich
Agarose, Special High EEO
Sigma-Aldrich
Agarose, Molecular Biology
Sigma-Aldrich
Agarose, Ultra-low Gelling Temperature
Sigma-Aldrich
Agarose, BioReagent, Molecular Biology, low EEO
Sigma-Aldrich
Agarose, High Gelling Temperature
Sigma-Aldrich
Agarose, Type IV, Special High EEO
Sigma-Aldrich
Sepharose 6B, 6% Beaded Agarose, 45-165 μm (wet), fractionation range 10,000-1,000,000 Da (dextrans)
Sigma-Aldrich
Agarose, Type I-A, low EEO
Sigma-Aldrich
Agarose, For pulsed field electrophoresis running gel