Merck
[Ethanol changes sensitivity of Kupffer cells to endotoxin].
Nihon Arukoru Yakubutsu Igakkai zasshi = Japanese journal of alcohol studies & drug dependence (2003-12-03)
Shunhei Yamashina, Kenichi Ikejima, Nobuyuki Enomoto, Yoshiyuki Takei, Nobuhiro Sato
ABSTRACT
Gut-derived endotoxin plays an important role in alcoholic liver injury. Intestinal sterilization with antibiotics (polymyxin B and neomycin) or inactivation of Kupffer cells with gadolinium chloride can prevent early alcohol-induced liver injury in the Tsukamoto-French model. Although short-term administration of alcohol enhances endotoxin hepatotoxicity, a majority of studies report that short-term ethanol inactivates Kupffer cells. It is therefore paradoxical that Kupffer cells are involved in alcoholic liver injury based on in vivo data with gadolinium chloride and antibiotics, yet ethanol blunts activation of isolated Kupffer cells. Accordingly, this review focuses on understanding this paradox by studying the temporal effect of ethanol in vivo on the response of subsequently isolated Kupffer cells. Mice were given ethanol intragastrically, and LPS was injected later. One hour after ethanol treatment, serum transaminases after LPS were 60% of control, while ethanol increased these parameters about 3-fold 21 hours after ethanol. Pretreatment with antibiotics blocked these effects of ethanol. Two hours after ethanol administration, the LPS-induced increases in intracellular calcium concentration and TNF alpha release by Kupffer cells was diminished by 50% of control, and these parameters were reciprocally enhanced two-fold at 24 hours. Sterilization of the gut with antibiotics blocked both effects of ethanol on intracellular calcium concentration and TNF alpha release. Twenty-four hours after ethanol, CD14 in Kupffer cells was elevated to about five-fold. In Kupffer cells from mice treated with ethanol 1 hour earlier, IRAK expression and activity and NF kappa B were decreased to 50-60% of control. In contrast, in Kupffer cells from mice treated with ethanol 21 hours earlier, LPS-induced TNF alpha production, expression and activity of IRAK were increased 1.5-fold over controls, while NF kappa B activation was elevated 3-fold. Kupffer cells isolated from rodents early after ethanol exhibited tolerance to LPS, whereas sensitization was observed later. In conclusion, acute ethanol alters the expression of endotoxin receptors and intracellular signaling molecules, and causes both tolerance and sensitization of Kupffer cells to endotoxin. It is postulated that tolerance of Kupffer cells contributes to the impairment of innate immune system in alcoholism, while sensitization to endotoxin enhances progression of alcoholic liver injury.
Sigma-Aldrich
Ethyl alcohol, Pure, 190 proof, ACS spectrophotometric grade, 95.0%
Supelco
Ethanol-10, 10 mg/dL in H2O, pack of 10 × 1.2 mL ampules, certified reference material, Cerilliant®
Supelco
Ethanol-150, 150 mg/dL in H2O, ampule of 10 × 1.2 mL, certified reference material, Cerilliant®
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, ACS reagent, ≥99.5%
Sigma-Aldrich
Ethyl alcohol, Pure 200 proof, Molecular Biology
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, anhydrous, ≥99.5%
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, meets USP testing specifications
Sigma-Aldrich
Ethanol
Sigma-Aldrich
Ethanol, BioUltra, Molecular Biology, ≥99.8%, (absolute alcohol, without additive, A15 o1)
Sigma-Aldrich
Ethanol, purum, fine spirit, denaturated with 4.8% methanol, F25 METHYL1, ~96% (based on denaturant-free substance)
Supelco
Ethanol, standard for GC
Sigma-Aldrich
Ethanol, purum, absolute ethanol, denaturated with 2% 2-butanone, A15 MEK1, ≥99.8% (based on denaturant-free substance)
Sigma-Aldrich
Ethanol, purum, absolute ethanol, denaturated with 4.8% isopropanol, A15 IPA1, ≥99.8% (based on denaturant-free substance)
Sigma-Aldrich
Ethanol, tested according to Ph. Eur.
Supelco
Ethanol-50, 50 mg/dL in H2O, ampule of 10 × 1.2 mL, certified reference material, Cerilliant®
Supelco
Ethanol-80, 80 mg/dL in H2O, ampule of 10 × 1.2 mL, certified reference material, Cerilliant®
Supelco
Ethanol-100 (10 ampules/kit), 100 mg/dL in H2O, ampule of 10 × 1.2 mL, certified reference material, Cerilliant®
Supelco
Ethanol-300, 300 mg/dL in H2O, ampule of 10 × 1.2 mL, certified reference material, Cerilliant®
Supelco
Ethanol-200 (10 ampules/kit), 200 mg/dL in H2O, ampule of 10 × 1.2 mL, certified reference material, Cerilliant®
Supelco
Ethanol solution, certified reference material, 2000 μg/mL in methanol
Sigma-Aldrich
Reagent Alcohol, anhydrous, ≤0.005% water
Sigma-Aldrich
Reagent Alcohol, anhydrous, ≤0.003% water
Sigma-Aldrich
Ethyl alcohol, Pure 190 proof, for molecular biology
Supelco
Dehydrated Alcohol, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Ethanol Calibration Kit, ampule of 10 × 1.2 mL, certified reference material, Cerilliant®
Supelco
Ethanol-400 (10 ampules/kit), 400 mg/dL in H2O, ampule of 10 × 1.2 mL, certified reference material, Cerilliant®
Supelco
Ethanol-500, 500 mg/dL in H2O, ampule of 10 × 1.2 mL, certified reference material, Cerilliant®
Supelco
Ethanol-20 (10 ampules/kit), 20 mg/dL in H2O, ampule of 10 × 1.2 mL, certified reference material, Cerilliant®