Merck
  • Ethanol-induced in vitro invasion of breast cancer cells: the contribution of MMP-2 by fibroblasts.
Ethanol-induced in vitro invasion of breast cancer cells: the contribution of MMP-2 by fibroblasts.
International journal of cancer (2004-09-24)
Moe Moe Aye, Cuiling Ma, Hong Lin, Kimberly A Bower, Richard C Wiggins, Jia Luo
ABSTRACT
Ethanol is a tumor promoter and may promote metastasis of breast cancer. However, the underlying cellular/molecular mechanisms remain unknown. Overexpression and high activity of matrix metalloproteinase-2 (MMP-2) are frequently associated with metastatic breast cancers and serve as a prognostic indicator of clinical outcome. MMP-2 is predominantly expressed in stromal fibroblasts and plays a pivotal role in regulating the invasive behavior of breast tumor cells. We hypothesized that ethanol may enhance the invasion of breast tumor cells by modulating the activity of fibroblastic MMP-2. With in vitro models (HS68 and CCD1056SK human fibroblasts), we showed that ethanol at physiologically relevant concentrations (50-200 mg/dl) activated MMP-2; conversely, at a higher concentration (400 mg/dl), it inhibited the MMP-2 activity. Consistently, conditioned medium collected from ethanol (50-200 mg/dl)-exposed fibroblasts markedly enhanced the invasive potential of breast cancer cells and mammary epithelial cells overexpressing ErbB2/HER2 (BT474, SKBR-3 and HB2(ErbB2) cells) but had little effect on cells with low ErbB2 levels (BT20 and HB2 cells). In contrast, conditioned medium obtained from ethanol (400 mg/dl)-treated fibroblasts inhibited cell invasion. Selective inhibitors of MMP-2 (SB-3CT and OA-Hy) eliminated ethanol-stimulated invasion, indicating that the effect of ethanol was mediated by MMP-2. Ethanol activated conventional PKCs and JNKs in fibroblasts; inhibitors of PKC (Go6850 and Go6976) and JNKs (SP600125) significantly inhibited ethanol-mediated MMP-2 activation as well as cell invasion, indicating that PKCs and JNKs play a role in ethanol-induced MMP-2 activation and cell invasion in vitro. Thus, ethanol-promoted breast cancer cell invasion may be mediated by the modulation of fibroblastic MMP-2.
Sigma-Aldrich
Reagent Alcohol, anhydrous, ≤0.003% water
Supelco
Ethanol solution, certified reference material, 2000 μg/mL in methanol
Sigma-Aldrich
Reagent Alcohol, anhydrous, ≤0.005% water
Supelco
Ethanol-400 (10 ampules/kit), 400 mg/dL in H2O, ampule of 10 × 1.2 mL, certified reference material, Cerilliant®
Supelco
Ethanol-500, 500 mg/dL in H2O, ampule of 10 × 1.2 mL, certified reference material, Cerilliant®
Supelco
Ethanol-20 (10 ampules/kit), 20 mg/dL in H2O, ampule of 10 × 1.2 mL, certified reference material, Cerilliant®
Sigma-Aldrich
Ethyl alcohol, Pure 190 proof, for molecular biology
Supelco
Dehydrated Alcohol, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Ethanol Calibration Kit, ampule of 10 × 1.2 mL, certified reference material, Cerilliant®
Supelco
Ethanol-10, 10 mg/dL in H2O, pack of 10 × 1.2 mL ampules, certified reference material, Cerilliant®
Supelco
Ethanol-150, 150 mg/dL in H2O, ampule of 10 × 1.2 mL, certified reference material, Cerilliant®
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, ACS reagent, ≥99.5%
Sigma-Aldrich
Ethanol, purum, absolute ethanol, denaturated with 2% 2-butanone, A15 MEK1, ≥99.8% (based on denaturant-free substance)
Sigma-Aldrich
Ethanol, purum, absolute ethanol, denaturated with 4.8% isopropanol, A15 IPA1, ≥99.8% (based on denaturant-free substance)
Sigma-Aldrich
Ethyl alcohol, Pure 200 proof, Molecular Biology
Sigma-Aldrich
Ethanol
Sigma-Aldrich
Ethyl alcohol, Pure, 190 proof, ACS spectrophotometric grade, 95.0%
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, anhydrous, ≥99.5%
Supelco
Ethanol, standard for GC
Sigma-Aldrich
Ethanol, tested according to Ph. Eur.
Sigma-Aldrich
Ethanol, purum, fine spirit, denaturated with 4.8% methanol, F25 METHYL1, ~96% (based on denaturant-free substance)
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, meets USP testing specifications
Sigma-Aldrich
Ethanol, BioUltra, Molecular Biology, ≥99.8%, (absolute alcohol, without additive, A15 o1)
Supelco
Ethanol-200 (10 ampules/kit), 200 mg/dL in H2O, ampule of 10 × 1.2 mL, certified reference material, Cerilliant®
Supelco
Ethanol-300, 300 mg/dL in H2O, ampule of 10 × 1.2 mL, certified reference material, Cerilliant®
Supelco
Ethanol-100 (10 ampules/kit), 100 mg/dL in H2O, ampule of 10 × 1.2 mL, certified reference material, Cerilliant®
Supelco
Ethanol-50, 50 mg/dL in H2O, ampule of 10 × 1.2 mL, certified reference material, Cerilliant®
Supelco
Ethanol-80, 80 mg/dL in H2O, ampule of 10 × 1.2 mL, certified reference material, Cerilliant®