Merck
  • Phytochemical constituents of Mongolian traditional medicinal plants, Chamaerhodos erecta and C. altaica, and its constituents prevents the extracellular matrix degradation factors.
Phytochemical constituents of Mongolian traditional medicinal plants, Chamaerhodos erecta and C. altaica, and its constituents prevents the extracellular matrix degradation factors.
Journal of natural medicines (2013-02-12)
Erdenechimeg Selenge, Gendaram Odontuya, Toshihiro Murata, Kenroh Sasaki, Kyoko Kobayashi, Javzan Batkhuu, Fumihiko Yoshizaki
ABSTRACT
Activity-guided isolation of the n-butanol fraction of Chamaerhodos erecta and water soluble fraction of C. altaica resulted in the isolation of 39 compounds, including new compounds identified as 4,5-dihydroxybenzaldehyde-3-O-β-D-glucopyranoside (1) from C. erecta and quercetin-3-O-β-D-glucuronopyranosyl-4'-O-β-D-glucopyranoside (2) from C. altaica. A total of 37 other compounds were identified based on physico-chemical properties and spectroscopic data. Antioxidative activity was evaluated using a DPPH radical-scavenging method, hyaluronidase inhibitory activity, and advanced glycation end products production inhibitory activity of isolated compounds. Some flavonols (4, 6, 9-11, 14, 15), catechins (18, 19), an amino acid (20), a lignan glucoside (23), and tannins (29-39) exhibited potential a free radical scavenging activity while the new compound (1) showed weak activity. A catechin (18) and some of the tannins (32, 33, 35, 36, 38) had moderate hyaluronidase inhibitory activity. Some of flavonoids and tannins prevented advanced glycation end products production, and the IC₅₀ of compounds 3, 9, 14-16, 33, 34, 36, 38, and 39 were determined.
Sigma-Aldrich
Hyaluronidase from sheep testes, Type V, lyophilized powder, ≥1,500 units/mg solid
Sigma-Aldrich
Hyaluronidase from sheep testes, Type II, lyophilized powder, ≥300 units/mg solid
Sigma-Aldrich
Hyaluronidase from bovine testes, Type IV-S, powder, suitable for mouse embryo cell culture, 750-3000 units/mg solid
Sigma-Aldrich
Hyaluronidase from bovine testes, Type VIII, lyophilized powder, 300-1,000 U/mg
Sigma-Aldrich
Hyaluronidase from bovine testes, Type IV-S, lyophilized powder (essentially salt-free), 750-3000 units/mg solid
Sigma-Aldrich
Hyaluronidase from bovine testes, Type I-S, lyophilized powder, 400-1000 units/mg solid
Sigma-Aldrich
Hyaluronidase from bovine testes, Type VI-S, lyophilized powder, 3,000-15,000 units/mg solid