Merck
Separation of ovotransferrin and ovomucoid from chicken egg white.
Poultry science (2014-04-08)
E D N S Abeyrathne, H Y Lee, D U Ahn
ABSTRACT
Ovotransferrin and ovomucoid were separated using 2 methods after extracting the ovotransferrin- and ovomucoid-containing fraction from egg white. Diluted egg white (2×) was added to Fe(3+) and treated with 43% ethanol (final concentration). After centrifugation, the supernatant was collected and treated with either a high-level ethanol (61% final concentration) or an acidic salt combination (2.5% ammonium sulfate and 2.5% citric acid) to separate ovotransferrin and ovomucoid. For the high-level of ethanol method, ovotransferrin was precipitated using 61% ethanol. After centrifugation, the precipitant was dissolved in 9 vol. of distilled water and the residual ethanol in the solution was removed using ultrafiltration. The supernatant, mainly containing ovomucoid, was diluted with 4 vol. of water, had ethanol removed, and was then concentrated and used as the ovomucoid fraction. For the acidic salt precipitation method, the ethanol in the supernatant was removed first. The ethanol-free solution was then concentrated and treated with a 2.5% ammonium sulfate and 2.5% citric acid combination. After centrifugation, the precipitant was used as the ovotransferrin and the supernatant as the ovomucoid fraction. The ovomucoid fraction from both of the protocols was further purified by heating at 65°C for 20 min and the impurities were removed by centrifugation. The yields of ovomucoid and ovotransferrin were >96 and >92%, respectively. The purity of ovomucoid was >89% and that of the ovotransferrin was >88%. The ELISA results confirmed that the activity of the separated ovotransferrin was >95%. Both of the protocols separated ovotransferrin and ovomucoid effectively and the methods were simple, fast, and easy to scale up.
Product Description
Supelco
Ethanol-10, 10 mg/dL in H2O, pack of 10 × 1.2 mL ampules, certified reference material, Cerilliant®
Supelco
Ethanol-150, 150 mg/dL in H2O, ampule of 10 × 1.2 mL, certified reference material, Cerilliant®
Sigma-Aldrich
Ethanol, purum, absolute ethanol, denaturated with 2% 2-butanone, A15 MEK1, ≥99.8% (based on denaturant-free substance)
Supelco
Ethanol, standard for GC
Sigma-Aldrich
Ethanol, tested according to Ph. Eur.
Sigma-Aldrich
Ethanol, purum, absolute ethanol, denaturated with 4.8% isopropanol, A15 IPA1, ≥99.8% (based on denaturant-free substance)
Sigma-Aldrich
Ethanol, BioUltra, Molecular Biology, ≥99.8%, (absolute alcohol, without additive, A15 o1)
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, anhydrous, ≥99.5%
Sigma-Aldrich
Ethanol
Sigma-Aldrich
Ethanol, purum, fine spirit, denaturated with 4.8% methanol, F25 METHYL1, ~96% (based on denaturant-free substance)
Sigma-Aldrich
Ethyl alcohol, Pure 200 proof, Molecular Biology
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, ACS reagent, ≥99.5%
Sigma-Aldrich
Ethyl alcohol, Pure, 190 proof, ACS spectrophotometric grade, 95.0%
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, meets USP testing specifications
Supelco
Ethanol-50, 50 mg/dL in H2O, ampule of 10 × 1.2 mL, certified reference material, Cerilliant®
Supelco
Ethanol-200 (10 ampules/kit), 200 mg/dL in H2O, ampule of 10 × 1.2 mL, certified reference material, Cerilliant®
Supelco
Ethanol-100 (10 ampules/kit), 100 mg/dL in H2O, ampule of 10 × 1.2 mL, certified reference material, Cerilliant®
Supelco
Ethanol-80, 80 mg/dL in H2O, ampule of 10 × 1.2 mL, certified reference material, Cerilliant®
Supelco
Ethanol-300, 300 mg/dL in H2O, ampule of 10 × 1.2 mL, certified reference material, Cerilliant®
Sigma-Aldrich
Reagent Alcohol, anhydrous, ≤0.005% water
Sigma-Aldrich
Reagent Alcohol, anhydrous, ≤0.003% water
Sigma-Aldrich
Ethyl alcohol, Pure 190 proof, for molecular biology
Supelco
Ethanol solution, certified reference material, 2000 μg/mL in methanol
Sigma-Aldrich
Conalbumin from chicken egg white, Iron complex
Sigma-Aldrich
Conalbumin from chicken egg white, suitable for cell culture, BioReagent
Sigma-Aldrich
Conalbumin from chicken egg white, Substantially iron-free
USP
Dehydrated Alcohol, United States Pharmacopeia (USP) Reference Standard
Supelco
Dehydrated Alcohol, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Ethanol-400 (10 ampules/kit), 400 mg/dL in H2O, ampule of 10 × 1.2 mL, certified reference material, Cerilliant®
Supelco
Ethanol-500, 500 mg/dL in H2O, ampule of 10 × 1.2 mL, certified reference material, Cerilliant®