Merck
  • Contribution of lipid environment to NADPH oxidase activity: influence of sterol.
Contribution of lipid environment to NADPH oxidase activity: influence of sterol.
Biochimie (2014-12-03)
Hager Souabni, Paul Machillot, Laura Baciou
ABSTRACT
The NADPH-oxidase complex, which plays beneficial or detrimental role in the inflammatory and degenerative diseases, is a membrane multi-subunit complex tightly regulated in order to produce superoxide anions, precursor of oxygen reactive species (ROS), in cells. The flavocytochrome b(558) (Cytb(558)) is the catalytic core of the NADPH oxidase which consists of two membrane proteins gp91(phox) (highly glycosylated) and p22(phox). In this work we took advantage of heterologous yeast cells engineered to express wild-type bovine Cytb(558) to analyze the properties of the NADPH oxidase activity during the biosynthesis processing steps of gp91(phox) and p22(phox) within endoplasmic reticulum (ER) and plasma membrane (Pmb). Our data showed that, in yeast, the heterodimerization at the endoplasmic reticulum membranes was concomitant with high level glycosylation of gp91(phox) and the heme acquisition. This study also demonstrated that the phagocyte NADPH oxidase was active at ER membranes and that this activity was surprisingly higher at the ER compared to the Pmb membranes. We have correlated these findings with the presence of sterols in the plasma membranes and their absence in ER membranes. This correlation was confirmed by decreased superoxide anion production rates in proteoliposomes supplemented with ergosterol or cholesterol. Our data support the idea that membrane environment might be determinant for ROS regulation and that sterols could directly interact with the membrane proteins of the NADPH oxidase constraining its capacity to produce superoxide anions.
Sigma-Aldrich
Arachidonic acid, from non-animal source, ≥98.5% (GC)
Sigma-Aldrich
Arachidonic acid, >95.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate solution, BioUltra, Molecular Biology, 10% in H2O
Sigma-Aldrich
Sodium dodecyl sulfate solution, BioUltra, 20% in H2O
Sodium laurilsulfate, European Pharmacopoeia (EP) Reference Standard
Supelco
Cholesterol, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Cholesterol, tested according to Ph. Eur.
Sigma-Aldrich
Cholesterol, from lanolin, ≥99.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, ≥98.0% (GC)
Supelco
Cholesterol solution, certified reference material, 10 mg/mL in chloroform
Sigma-Aldrich
SyntheChol® NS0 Supplement, 500 ×, synthetic cholesterol, animal component-free, sterile-filtered, aqueous solution, suitable for cell culture
Sigma-Aldrich
1,2-Dimyristoyl-sn-glycero-3-phosphocholine, ≥99%
Sigma-Aldrich
Cholesterol, powder, BioReagent, suitable for cell culture, ≥99%
Sigma-Aldrich
Cholesterol, Sigma Grade, ≥99%
SAFC
Cholesterol, from sheep wool, Controlled origin, meets USP/NF testing specifications
Sigma-Aldrich
Sodium dodecyl sulfate, BioReagent, Molecular Biology, ≥98.5% (GC), free-flowing, Redi-Dri
Sigma-Aldrich
Sodium dodecyl sulfate, ≥98.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, BioXtra, ≥99.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, 92.5-100.5% based on total alkyl sulfate content basis
Supelco
Sodium dodecyl sulfate, suitable for ion pair chromatography, LiChropur, ≥99.0%
Supelco
Sodium dodecyl sulfate, dust-free pellets, suitable for electrophoresis, Molecular Biology, ≥99.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, BioReagent, suitable for electrophoresis, Molecular Biology, ≥98.5% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, ACS reagent, ≥99.0%
Sigma-Aldrich
Sodium dodecyl sulfate, ≥99.0% (GC), dust-free pellets
Sigma-Aldrich
Sodium dodecyl sulfate, ≥90% ((Assay))
Sigma-Aldrich
Sodium dodecyl sulfate, tested according to NF, mixture of sodium alkyl sulfates consisting mainly of sodium dodecyl sulfate
Sigma-Aldrich
Sodium dodecyl sulfate, BioUltra, Molecular Biology, ≥99.0% (GC)
Sigma-Aldrich
Cholesterol, from sheep wool, ≥92.5% (GC), powder
Sigma-Aldrich
Sodium dodecyl sulfate, ReagentPlus®, ≥98.5% (GC)