Merck
  • Methyltransferase-like 17 physically and functionally interacts with estrogen receptors.
Methyltransferase-like 17 physically and functionally interacts with estrogen receptors.
IUBMB life (2015-10-22)
Peiyun Du, Bin Yuan, Jia Cao, Jing Zhao, Lihua Ding, Lihan Chen, Sunyang Ying, Lina Jiang, Jiajia Lin, Xiaojie Xu, Long Cheng, Qinong Ye
ABSTRACT
Estrogen exerts its physiological and pathological functions through two estrogen receptors (ERs), ERα and ERβ, which act as transcription factors. Coregulators, including coactivators and corepressors, have been shown to be crucial for regulation of ER transcriptional activity. Although many coregulators have been identified to regulate activities of ERs, novel coregulators are still needed to be investigated. Here, we show that human methyltransferase-like 17 (METTL17), whose function is unknown, physically interacts with ERα and ERβ, and functionally acts as a coactivator for ERs. METTL17 interacts with ER in vitro and in yeast and mammalian cells. Activation function-1 (AF1) and AF2 domains of ERs are responsible for the interaction between METTL17 and ERs. Knockdown of METTL17 reduces transcriptional activities of ERα and ERβ in breast cancer cells, whereas METTL17 overexpression increases ERα and ERβ transcriptional activities. Inhibition of METTL17 expression decreases mRNA and protein levels of ER target genes, including PR, cathepsin D, and pS2. Moreover, METTL17 knockdown reduces breast cancer cell growth. These results indicate that METTL17 is a novel coactivator of ERs and may play a role in breast tumorigenesis.
Sigma-Aldrich
Sodium dodecyl sulfate, ≥98.0% (GC)
Sigma-Aldrich
DL-Dithiothreitol solution, BioUltra, Molecular Biology, ~1 M in H2O
Sigma-Aldrich
Sodium dodecyl sulfate, ≥90% ((Assay))
Millipore
Anti-c-Myc Agarose Affinity Gel antibody produced in rabbit, affinity isolated antibody
Sigma-Aldrich
Sodium chloride, AnhydroBeads, −10 mesh, 99.999% trace metals basis
Sigma-Aldrich
Sodium chloride, BioUltra, Molecular Biology, ≥99.5% (AT)
Sigma-Aldrich
Sodium dodecyl sulfate, BioUltra, Molecular Biology, ≥99.0% (GC)
Sigma-Aldrich
Phenylmethanesulfonyl fluoride, ≥99.0% (T)
Sigma-Aldrich
Sodium dodecyl sulfate, ReagentPlus®, ≥98.5% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, ≥99.0% (GC), dust-free pellets
Sigma-Aldrich
Sodium chloride, 99.999% trace metals basis
Sigma-Aldrich
Sodium chloride solution, 0.85%
Sigma-Aldrich
Sodium dodecyl sulfate, ACS reagent, ≥99.0%
Sigma-Aldrich
MISSION® esiRNA, targeting mouse Mett11d1
Sigma-Aldrich
Sodium dodecyl sulfate, tested according to NF, mixture of sodium alkyl sulfates consisting mainly of sodium dodecyl sulfate
Sigma-Aldrich
Sodium chloride solution, BioUltra, Molecular Biology, ~5 M in H2O
Sigma-Aldrich
Sodium dodecyl sulfate, BioXtra, ≥99.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, 92.5-100.5% based on total alkyl sulfate content basis
Sigma-Aldrich
Sodium dodecyl sulfate, BioReagent, Molecular Biology, ≥98.5% (GC)
Sigma-Aldrich
Sodium chloride, BioXtra, ≥99.5% (AT)
Sigma-Aldrich
Sodium chloride solution, 0.9% in water, BioXtra, suitable for cell culture
Sigma-Aldrich
Sodium chloride, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99%
Sigma-Aldrich
Sodium chloride, tablet
Sigma-Aldrich
Sodium chloride, Molecular Biology, DNase, RNase, and protease, none detected, ≥99% (titration)
Sigma-Aldrich
Phenylmethanesulfonyl fluoride, ≥98.5% (GC)
Sigma-Aldrich
Sodium chloride solution, 5 M in H2O, BioReagent, Molecular Biology
Sigma-Aldrich
Sodium chloride, meets analytical specification of Ph. Eur., BP, USP, 99.0-100.5%
Sigma-Aldrich
Sodium chloride solution, 5 M
Sigma-Aldrich
Sodium chloride-35Cl, 99 atom % 35Cl
Sigma-Aldrich
Sodium chloride, random crystals, 99.9% trace metals basis