Merck
  • An integrated genome-wide approach to discover deregulated microRNAs in non-small cell lung cancer: Clinical significance of miR-23b-3p deregulation.
An integrated genome-wide approach to discover deregulated microRNAs in non-small cell lung cancer: Clinical significance of miR-23b-3p deregulation.
Scientific reports (2015-09-01)
Shahnaz Begum, Masamichi Hayashi, Takenori Ogawa, Fayez J Jabboure, Mariana Brait, Evgeny Izumchenko, Sarit Tabak, Steven A Ahrendt, William H Westra, Wayne Koch, David Sidransky, Mohammad O Hoque
ABSTRACT
In spite of significant technical advances, genesis and progression of non-small cell lung cancer (NSCLC) remain poorly understood. We undertook an integrated genetic approach to discover novel microRNAs that were deregulated in NSCLCs. A total 119 primary NSCLCs with matched normal were analyzed for genome-wide copy number changes. We also tested a subset of matched samples by microRNA expression array, and integrated them to identify microRNAs positioned in allelic imbalance area. Our findings support that most of the identified deregulated microRNAs (miR-21, miR-23b, miR-31, miR-126, miR-150, and miR-205) were positioned in allelic imbalance areas. Among microRNAs tested in independent 114 NSCLCs, overexpression of miR-23b was revealed to be a significantly poor prognostic factor of recurrence free survival (HR = 2.40, P = 0.005, 95%CI: 1.32-4.29) and overall survival (HR = 2.35, P = 0.005, 95%CI: 1.30-4.19) in multivariable analysis. In addition, overexpression of miR-23b in H1838 cell line significantly increased cell proliferation, while inhibition of miR-23b in H1437 and H1944 cell lines significantly decreased cell doubling time. In summary, integration of genomic analysis and microRNA expression profiling could identify novel cancer-related microRNAs, and miR-23b could be a potential prognostic marker for early stage NSCLCs. Further biological studies of miR-23b are warranted for the potential development of targeted therapy.
Sigma-Aldrich
Dimethyl sulfoxide, Hybri-Max, sterile-filtered, BioReagent, suitable for hybridoma, ≥99.7%
Sigma-Aldrich
Dimethyl sulfoxide, meets EP testing specifications, meets USP testing specifications
Sigma-Aldrich
8-Octanoyloxypyrene-1,3,6-trisulfonic acid trisodium salt, suitable for fluorescence, ≥90% (HPCE)
Sigma-Aldrich
Dimethyl sulfoxide, anhydrous, ≥99.9%
Sigma-Aldrich
Sodium dodecyl sulfate solution, BioUltra, Molecular Biology, 10% in H2O
Sigma-Aldrich
Sodium dodecyl sulfate solution, BioUltra, 20% in H2O
Sigma-Aldrich
Dimethyl sulfoxide, BioUltra, Molecular Biology, ≥99.5% (GC)
Sigma-Aldrich
Dimethyl sulfoxide, ≥99.5% (GC), suitable for plant cell culture
Sigma-Aldrich
Dimethyl sulfoxide, sterile-filtered, BioPerformance Certified, meets EP, USP testing specifications, suitable for hybridoma
Sigma-Aldrich
Dimethyl sulfoxide, Molecular Biology
Sigma-Aldrich
Dimethyl sulfoxide, PCR Reagent
Sigma-Aldrich
Dimethyl sulfoxide, ≥99.6%, ReagentPlus®
Sigma-Aldrich
L-Thyroxine sodium salt pentahydrate, ≥98% (HPLC), powder
Sigma-Aldrich
Thiazolyl Blue Tetrazolium Bromide, powder, BioReagent, suitable for cell culture, suitable for insect cell culture, ≥97.5% (HPLC)
Sigma-Aldrich
L-Thyroxine sodium salt pentahydrate, γ-irradiated, powder, BioXtra, suitable for cell culture
Sigma-Aldrich
Thiazolyl Blue Tetrazolium Bromide, 98%
Sigma-Aldrich
Nitrogen-14N2, 99.99 atom % 14N
Sigma-Aldrich
Sodium dodecyl sulfate, BioReagent, Molecular Biology, ≥98.5% (GC), free-flowing, Redi-Dri
Sigma-Aldrich
Sodium dodecyl sulfate, ≥98.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, BioXtra, ≥99.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, 92.5-100.5% based on total alkyl sulfate content basis
Supelco
Sodium dodecyl sulfate, dust-free pellets, suitable for electrophoresis, Molecular Biology, ≥99.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, BioReagent, suitable for electrophoresis, Molecular Biology, ≥98.5% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, ACS reagent, ≥99.0%
Sigma-Aldrich
Sodium dodecyl sulfate, ≥99.0% (GC), dust-free pellets
Sigma-Aldrich
Sodium dodecyl sulfate, ≥90% ((Assay))
Sigma-Aldrich
Sodium dodecyl sulfate, tested according to NF, mixture of sodium alkyl sulfates consisting mainly of sodium dodecyl sulfate
Sigma-Aldrich
Sodium dodecyl sulfate, BioUltra, Molecular Biology, ≥99.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, ReagentPlus®, ≥98.5% (GC)