Merck
  • Molecular and biochemical characterisation of human short-chain dehydrogenase/reductase member 3 (DHRS3).
Molecular and biochemical characterisation of human short-chain dehydrogenase/reductase member 3 (DHRS3).
Chemico-biological interactions (2014-12-03)
Tereza Lundová, Lucie Zemanová, Beata Malčeková, Adam Skarka, Hana Štambergová, Jana Havránková, Miroslav Šafr, Vladimír Wsól
ABSTRACT
Dehydrogenase/reductase (SDR family) member 3 (DHRS3), also known as retinal short-chain dehydrogenase/reductase (retSDR1) is a member of SDR16C family. This family is thought to be NADP(H) dependent and to have multiple substrates; however, to date, only all-trans-retinal has been identified as a DHRS3 substrate. The reductive reaction catalysed by DHRS3 seems to be physiological, and recent studies proved the importance of DHRS3 for maintaining suitable retinoic acid levels during embryonic development in vivo. Although it seems that DHRS3 is an important protein, knowledge of the protein and its properties is quite limited, with the majority of information being more than 15 years old. This study aimed to generate a more comprehensive characterisation of the DHRS3 protein. Recombinant enzyme was prepared and demonstrated to be a microsomal, integral-membrane protein with the C-terminus oriented towards the cytosol, consistent with its preference of NADPH as a cofactor. It was determined that DHRS3 also participates in the metabolism of other endogenous compounds, such as androstenedione, estrone, and DL-glyceraldehyde, and in the biotransformation of xenobiotics (e.g., NNK and acetohexamide) in addition to all-trans-retinal. Purified and reconstituted enzyme was prepared for the first time and will be used for further studies. Expression of DHRS3 was shown at the level of both mRNA and protein in the human liver, testis and small intestine. This new information could open other areas of DHRS3 protein research.
Sigma-Aldrich
Sodium dodecyl sulfate, ≥99.0% (GC), dust-free pellets
Sigma-Aldrich
Ammonia, anhydrous, ≥99.98%
Sigma-Aldrich
Sodium dodecyl sulfate, ACS reagent, ≥99.0%
Sigma-Aldrich
Ammonia solution, 0.4 M in THF
Sigma-Aldrich
Phenylmethanesulfonyl fluoride, ≥99.0% (T)
Sigma-Aldrich
Sodium dodecyl sulfate, tested according to NF, mixture of sodium alkyl sulfates consisting mainly of sodium dodecyl sulfate
Sigma-Aldrich
Sodium dodecyl sulfate, ≥90% ((Assay))
Sigma-Aldrich
Sodium dodecyl sulfate, BioUltra, Molecular Biology, ≥99.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, ≥98.0% (GC)
Sigma-Aldrich
n-Dodecyl β-D-maltoside, ≥98% (GC)
Sigma-Aldrich
n-Dodecyl β-D-maltoside, BioXtra, ≥98% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, BioXtra, ≥99.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, 92.5-100.5% based on total alkyl sulfate content basis
Sigma-Aldrich
L-Thyroxine sodium salt pentahydrate, ≥98% (HPLC), powder
Sigma-Aldrich
Sodium dodecyl sulfate, BioReagent, Molecular Biology, ≥98.5% (GC)
Sigma-Aldrich
Phenylmethanesulfonyl fluoride, ≥98.5% (GC)
Sigma-Aldrich
Potassium phosphate tribasic, reagent grade, ≥97%
Sigma-Aldrich
1,2-Dipalmitoyl-sn-glycero-3-phosphoethanolamine, ≥97%
Sigma-Aldrich
Sodium dodecyl sulfate, ReagentPlus®, ≥98.5% (GC)
Sigma-Aldrich
L-Thyroxine sodium salt pentahydrate, γ-irradiated, powder, BioXtra, suitable for cell culture
Supelco
Sodium dodecyl sulfate, dust-free pellets, suitable for electrophoresis, Molecular Biology, ≥99.0% (GC)
Sigma-Aldrich
Ammonia-14N, 99.99 atom % 14N
Sigma-Aldrich
Ammonia solution, 4 M in methanol
Sigma-Aldrich
Sodium dodecyl sulfate, BioReagent, Molecular Biology, ≥98.5% (GC), free-flowing, Redi-Dri
Sigma-Aldrich
Ethyl acetate, ≥99%, FCC, FG
Sigma-Aldrich
Ethyl acetate, natural, ≥99%, FCC, FG
Sigma-Aldrich
Sucrose, puriss., meets analytical specification of Ph. Eur., BP, NF
Sigma-Aldrich
Sucrose, BioUltra, Molecular Biology, ≥99.5% (HPLC)
Sigma-Aldrich
Phenylacetic acid, ≥99%, FCC, FG
Sigma-Aldrich
Sodium dodecyl sulfate solution, BioUltra, 20% in H2O