Skip to Content
Merck

BDPH9BCC39EF

Bide Pharmatech

Ethyl 6-(aminomethyl)nicotinate

Synonym(s):

ethyl 6-(aminomethyl)nicotinate

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Linear Formula:
C9H12O2N2
CAS Number:
Molecular Weight:
180.21
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Inchi Code

1S/C9H12N2O2/c1-2-13-9(12)7-3-4-8(5-10)11-6-7/h3-4,6H,2,5,10H2,1H3

Inchi Key

YPRBNFRLIKIHST-UHFFFAOYSA-N

Shipping Temperature

Normal

Purity

97%

Storage temp.

Keep in dark place, inert atmosphere, 2-8C

Country of Origin

CN

Physical Form

Powder or crystals



Still not finding the right product?

Explore all of our products under Ethyl 6-(aminomethyl)nicotinate




Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library