Skip to Content
Merck

BDPH9BCC4CD5

Bide Pharmatech

2,3-Difluoro-6-methoxybenzoic acid

Synonym(s):

2,3-difluoro-6-methoxybenzoic acid

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Linear Formula:
C8H6O3F2
CAS Number:
Molecular Weight:
188.13
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Inchi Code

1S/C8H6F2O3/c1-13-5-3-2-4(9)7(10)6(5)8(11)12/h2-3H,1H3,(H,11,12)

Inchi Key

MMTKYWQMCSZCGW-UHFFFAOYSA-N

Shipping Temperature

Normal

Purity

98%

Storage temp.

Sealed in dry, room temperature

Country of Origin

CN

Physical Form

Solid



Still not finding the right product?

Explore all of our products under 2,3-Difluoro-6-methoxybenzoic acid


hstatements

Precautionary Statements

Signal Word

Warning

Pictograms

Exclamation mark


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library