Skip to Content
Merck

BDPH9BCC9D67

Bide Pharmatech

Dimethyl 2,3,5,6-tetrachloroterephthalate

Synonym(s):

dimethyl 2,3,5,6-tetrachloroterephthalate

Select a Size

Change View

About This Item

Linear Formula:
C10H6O4Cl4
CAS Number:
Molecular Weight:
331.97
MDL number:
Sign in to see your pricing
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Inchi Code

1S/C10H6Cl4O4/c1-17-9(15)3-5(11)7(13)4(10(16)18-2)8(14)6(3)12/h1-2H3

Inchi Key

NPOJQCVWMSKXDN-UHFFFAOYSA-N

Shipping Temperature

Normal

Purity

96%

Storage temp.

Sealed in dry, room temperature

Country of Origin

CN

Physical Form

Solid



hstatements

Precautionary Statements

Signal Word

Warning

Pictograms

Exclamation mark


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library