Skip to Content
Merck

BDPH9BCC9DA2

Bide Pharmatech

3-Chloro-4-(pyridin-3-yloxy)aniline

Synonym(s):

3-chloro-4-(3-pyridinyloxy)aniline

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Linear Formula:
C11H9O1N2Cl1
CAS Number:
Molecular Weight:
220.66
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Inchi Code

1S/C11H9ClN2O/c12-10-6-8(13)3-4-11(10)15-9-2-1-5-14-7-9/h1-7H,13H2

Inchi Key

HUHJCGBQPHGZAW-UHFFFAOYSA-N

Shipping Temperature

Normal

Purity

98%

Storage temp.

Keep in dark place, inert atmosphere, room temperature

Country of Origin

CN

Physical Form

Solid



Still not finding the right product?


hstatements

Signal Word

Warning

Pictograms

Exclamation mark


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library