Skip to Content
Merck

BDPH9BCCB7F4

Bide Pharmatech

tert-Butyl (3-(oxiran-2-yl)propyl)carbamate

Synonym(s):

tert-butyl 3-(oxiran-2-yl)propylcarbamate

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Linear Formula:
C10H19O3N1
CAS Number:
Molecular Weight:
201.27
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Inchi Code

1S/C10H19NO3/c1-10(2,3)14-9(12)11-6-4-5-8-7-13-8/h8H,4-7H2,1-3H3,(H,11,12)

Inchi Key

WAUPLTBYJNZQJL-UHFFFAOYSA-N

Shipping Temperature

Cold-chain transportation

Purity

95%

Storage temp.

Sealed in dry, store in freezer, under -20C

Country of Origin

CN

Physical Form

Liquid or semi-solid



Still not finding the right product?



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library