Skip to Content

BDPH9BCD0D0F

Bide Pharmatech

4-(Trifluoromethyl)quinoline

Synonym(s):

4-(trifluoromethyl)quinoline

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Linear Formula:
C10H6N1F3
CAS Number:
Molecular Weight:
197.16
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Inchi Code

1S/C10H6F3N/c11-10(12,13)8-5-6-14-9-4-2-1-3-7(8)9/h1-6H

Inchi Key

CLYXBXQWMBMXLO-UHFFFAOYSA-N

Shipping Temperature

Normal

Purity

97%

Storage temp.

Sealed in dry, 2-8C

Country of Origin

CN

Physical Form

Solid or liquid



Still not finding the right product?

Explore all of our products under 4-(Trifluoromethyl)quinoline


Precautionary Statements

Signal Word

Warning

Pictograms

Exclamation mark


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library