Skip to Content
Merck

BDPH9BCD24CC

Bide Pharmatech

6-Bromopyrazolo[1,5-a]pyridine

Synonym(s):

6-bromopyrazolo[1,5-a]pyridine

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Linear Formula:
C7H5N2Br1
CAS Number:
Molecular Weight:
197.03
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Inchi Code

1S/C7H5BrN2/c8-6-1-2-7-3-4-9-10(7)5-6/h1-5H

Inchi Key

MJGAKVFFADUFMG-UHFFFAOYSA-N

Shipping Temperature

Normal

Purity

97%

Storage temp.

Sealed in dry, room temperature

Country of Origin

CN

Physical Form

Solid or liquid



Still not finding the right product?

Explore all of our products under 6-Bromopyrazolo[1,5-a]pyridine


Precautionary Statements

Signal Word

Warning

Pictograms

Exclamation mark


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library