Skip to Content

BDPH9BCD2B26

Bide Pharmatech

5-Bromo-N-hydroxypyridine-2-carboxamide

Synonym(s):

5-bromo-N-hydroxy-2-pyridinecarboxamide

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Linear Formula:
C6H5O2N2Br1
CAS Number:
Molecular Weight:
217.02
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Inchi Code

1S/C6H5BrN2O2/c7-4-1-2-5(8-3-4)6(10)9-11/h1-3,11H,(H,9,10)

Inchi Key

OUJLLBUAKZPKEY-UHFFFAOYSA-N

Shipping Temperature

Normal

Purity

95%

Storage temp.

Inert atmosphere, room temperature

Country of Origin

CN

Physical Form

Powder or crystals



Still not finding the right product?


Pictograms

Flame

hstatements

Precautionary Statements

Signal Word

Danger



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library