Skip to Content

BDPH9BCD2F62

Bide Pharmatech

6-Chloro-3-fluoropyridin-2-amine

Synonym(s):

6-chloro-3-fluoro-2-pyridinamine

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Linear Formula:
C5H4N2Cl1F1
CAS Number:
Molecular Weight:
146.55
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Inchi Code

1S/C5H4ClFN2/c6-4-2-1-3(7)5(8)9-4/h1-2H,(H2,8,9)

Inchi Key

WXZISDKNBACWRH-UHFFFAOYSA-N

Shipping Temperature

Normal

Purity

97%

Storage temp.

Keep in dark place, inert atmosphere, room temperature

Country of Origin

CN

Physical Form

White to off-white to brown powder or crystals



Still not finding the right product?

Explore all of our products under 6-Chloro-3-fluoropyridin-2-amine


Precautionary Statements

Signal Word

Warning

Pictograms

Exclamation mark


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library