Skip to Content

BDPH9BCD4F8E

Bide Pharmatech

2-(Cyclopropyl(1-(thiazol-2-yl)ethyl)amino)acetic acid

Synonym(s):

{cyclopropyl[1-(1,3-thiazol-2-yl)ethyl]amino}acetic acid

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Linear Formula:
C10H14O2N2S1
CAS Number:
Molecular Weight:
226.3
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Inchi Code

1S/C10H14N2O2S/c1-7(10-11-4-5-15-10)12(6-9(13)14)8-2-3-8/h4-5,7-8H,2-3,6H2,1H3,(H,13,14)

Inchi Key

QJUVLUUNSHTPKO-UHFFFAOYSA-N

Shipping Temperature

Normal

Purity

97%

Storage temp.

Sealed in dry, 2-8C

Country of Origin

CN

Physical Form

Solid or semi-solid or liquid



Still not finding the right product?




Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library