Skip to Content
Merck

BDPH9BCD53F3

Bide Pharmatech

Dodecyl alcoho

Synonym(s):

1-dodecanol

Select a Size

Change View

About This Item

Linear Formula:
C12H26O1
CAS Number:
Molecular Weight:
186.34
MDL number:
Sign in to see your pricing
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Inchi Code

1S/C12H26O/c1-2-3-4-5-6-7-8-9-10-11-12-13/h13H,2-12H2,1H3

Inchi Key

LQZZUXJYWNFBMV-UHFFFAOYSA-N

Shipping Temperature

Normal

Purity

99%

Storage temp.

Sealed in dry, room temperature

Country of Origin

CN

Physical Form

Liquid or Semi-Solid or Solid or Lump



Still not finding the right product?

Explore all of our products under Dodecyl alcoho




Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library