Inchi Code
1S/C9H9Br/c1-7(2)8-4-3-5-9(10)6-8/h3-6H,1H2,2H3
Inchi Key
BHFSXGLQLQHDCH-UHFFFAOYSA-N
Shipping Temperature
Cold-chain transportation
Purity
98%
Storage temp.
Inert atmosphere, 2-8C
Country of Origin
CN
Physical Form
Liquid
Still not finding the right product?
Explore all of our products under 1-Bromo-3-(prop-1-en-2-yl)benzene
hstatements
Precautionary Statements
Signal Word
Warning
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
