Sign In to View Organizational & Contract Pricing.
Select a Size
Change View
| Size/SKU | Availability | Price |
|---|
About This Item
Pricing and availability is not currently available. Contact Technical Service
Inchi Code
1S/C8H8BrNO2/c1-12-8(11)4-6-2-3-10-5-7(6)9/h2-3,5H,4H2,1H3
Inchi Key
AMMJWCPUBIBZEG-UHFFFAOYSA-N
Shipping Temperature
Normal
Purity
97%
Country of Origin
CN
Physical Form
Liquid
Storage temp.
Inert atmosphere, 2-8°C
Still not finding the right product?
Explore all of our products under Methyl 2-(3-bromopyridin-4-yl)acetate
hstatements
Signal Word
Warning
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
