Skip to Content
Merck

BDPH9BCD8CF8

Bide Pharmatech

Methyl 3-acetyl-4-fluorobenzoate

Synonym(s):

methyl 3-acetyl-4-fluorobenzoate

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Linear Formula:
C10H9O3F1
CAS Number:
Molecular Weight:
196.18
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Inchi Code

1S/C10H9FO3/c1-6(12)8-5-7(10(13)14-2)3-4-9(8)11/h3-5H,1-2H3

Inchi Key

UUACGIJGQLBVJL-UHFFFAOYSA-N

Shipping Temperature

Normal

Purity

98%

Storage temp.

Sealed in dry, room temperature

Country of Origin

CN

Physical Form

Liquid



Still not finding the right product?

Explore all of our products under Methyl 3-acetyl-4-fluorobenzoate


Precautionary Statements

Signal Word

Warning

Pictograms

Exclamation mark


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library