Skip to Content
Merck

BDPH9BCD93D7

Bide Pharmatech

2-Chloro-5-(trifluoromethoxy)isonicotinic acid

Synonym(s):

2-chloro-5-(trifluoromethoxy)isonicotinic acid

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Linear Formula:
C7H3O3N1Cl1F3
CAS Number:
Molecular Weight:
241.55
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Inchi Code

1S/C7H3ClF3NO3/c8-5-1-3(6(13)14)4(2-12-5)15-7(9,10)11/h1-2H,(H,13,14)

Inchi Key

MXTVKRCSGPEYAX-UHFFFAOYSA-N

Shipping Temperature

Normal

Purity

95%

Storage temp.

Sealed in dry, 2-8C

Country of Origin

CN

Physical Form

White to Yellow Solid



Still not finding the right product?



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library