Sign In to View Organizational & Contract Pricing.
Select a Size
Change View
| Size/SKU | Availability | Price |
|---|
About This Item
Linear Formula:
C5H2O2N1Cl1Br1S1F1
CAS Number:
Molecular Weight:
274.5
MDL number:
Pricing and availability is not currently available. Contact Technical Service
Inchi Code
1S/C5H2BrClFNO2S/c6-4-2-1-3(5(8)9-4)12(7,10)11/h1-2H
Inchi Key
VADHBJNKHDTBMX-UHFFFAOYSA-N
Shipping Temperature
Normal
Purity
95%
Storage temp.
Inert atmosphere, 2-8C
Country of Origin
CN
Physical Form
Powder or crystals
Still not finding the right product?
Explore all of our products under 6-Bromo-2-fluoropyridine-3-sulfonyl chloride
hstatements
Precautionary Statements
Signal Word
Danger
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.

