Skip to Content
Merck

BDPH9BCDC8CF

Bide Pharmatech

6-Ethoxybenzo[d]oxazol-2-amine

Synonym(s):

6-ethoxy-1,3-benzoxazol-2-amine

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Linear Formula:
C9H10O2N2
CAS Number:
Molecular Weight:
178.19
MDL number:
Product BDPH9BCDC8CF is not currently sold in your country.


Inchi Code

1S/C9H10N2O2/c1-2-12-6-3-4-7-8(5-6)13-9(10)11-7/h3-5H,2H2,1H3,(H2,10,11)

Inchi Key

LAYYUBFRHUZDKA-UHFFFAOYSA-N

Shipping Temperature

Normal

Purity

95%

Storage temp.

Keep in dark place, inert atmosphere, room temperature

Country of Origin

CN

Physical Form

Solid



Still not finding the right product?

Explore all of our products under 6-Ethoxybenzo[d]oxazol-2-amine



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library