Skip to Content

BDPH9BCDCED1

Bide Pharmatech

1H-Imidazole-2-carboxamide

Synonym(s):

1H-imidazole-2-carboxamide

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Linear Formula:
C4H5O1N3
CAS Number:
Molecular Weight:
111.1
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Inchi Code

1S/C4H5N3O/c5-3(8)4-6-1-2-7-4/h1-2H,(H2,5,8)(H,6,7)

Inchi Key

NMIZONYLXCOHEF-UHFFFAOYSA-N

Shipping Temperature

Normal

Purity

97%

Storage temp.

Sealed in dry, room temperature

Country of Origin

CN

Physical Form

White to Yellow Solid



Still not finding the right product?

Explore all of our products under 1H-Imidazole-2-carboxamide


Precautionary Statements

Signal Word

Warning

Pictograms

Exclamation mark


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library