Skip to Content
Merck

BDPH9BCDD60E

Bide Pharmatech

2-(Dimethylamino)benzaldehyde

Synonym(s):

2-(dimethylamino)benzaldehyde

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Linear Formula:
C9H11O1N1
CAS Number:
Molecular Weight:
149.19
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Inchi Code

1S/C9H11NO/c1-10(2)9-6-4-3-5-8(9)7-11/h3-7H,1-2H3

Inchi Key

DGPBVJWCIDNDPN-UHFFFAOYSA-N

Shipping Temperature

Normal

Purity

98%

Storage temp.

Inert atmosphere, 2-8C

Country of Origin

CN

Physical Form

Colorless to light-yellow to yellow liquid



Still not finding the right product?

Explore all of our products under 2-(Dimethylamino)benzaldehyde


hstatements

Signal Word

Warning

Pictograms

Exclamation mark


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library