Skip to Content
Merck

BDPH9BCDE72E

Bide Pharmatech

(2,5-Dibromophenyl)hydrazine

Synonym(s):

1-(2,5-dibromophenyl)hydrazine

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Linear Formula:
C6H6N2Br2
CAS Number:
Molecular Weight:
265.94
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Inchi Code

1S/C6H6Br2N2/c7-4-1-2-5(8)6(3-4)10-9/h1-3,10H,9H2

Inchi Key

MVYLGBZJXJXCPY-UHFFFAOYSA-N

Shipping Temperature

Shipped in cold pack

Purity

97%

Storage temp.

Inert atmosphere, store in freezer, under -20C

Country of Origin

CN

Physical Form

White to Yellow Solid



Still not finding the right product?

Explore all of our products under (2,5-Dibromophenyl)hydrazine


Precautionary Statements

Signal Word

Warning

Pictograms

Exclamation mark


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library