Skip to Content
Merck

BDPH9BCDFA38

Bide Pharmatech

1-(1H-Imidazol-2-yl)-2-methylpropan-1-amine

Synonym(s):

1-(1H-imidazol-2-yl)-2-methyl-1-propanamine

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Linear Formula:
C7H13N3
CAS Number:
Molecular Weight:
139.2
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Inchi Code

1S/C7H13N3/c1-5(2)6(8)7-9-3-4-10-7/h3-6H,8H2,1-2H3,(H,9,10)

Inchi Key

WBNFIORQLJOELA-UHFFFAOYSA-N

Shipping Temperature

Normal

Purity

95%

Storage temp.

Keep in dark place, inert atmosphere, 2-8C

Country of Origin

CN

Physical Form

Solid or semi-solid or lump or liquid



Still not finding the right product?



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library