Skip to Content
Merck

BDPH9C021D63

Bide Pharmatech

(2S,4R)-1-((S)-3,3-Dimethyl-2-(pent-4-ynamido)butanoyl)-4-hydroxy-N-(4-(4-methylthiazol-5-yl)benzyl)pyrrolidine-2-carboxamide

Synonym(s):

(2S,4R)-4-hydroxy-1-((S)-2-((1-hydroxypent-4-yn-1-ylidene)amino)-3,3-dimethylbutanoyl)-N-(4-(4-methylthiazol-5-yl)benzyl)pyrrolidine-2-carbimidic acid

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

Product BDPH9C021D63 is not currently sold in your country.


Inchi Code

1S/C27H34N4O4S/c1-6-7-8-22(33)30-24(27(3,4)5)26(35)31-15-20(32)13-21(31)25(34)28-14-18-9-11-19(12-10-18)23-17(2)29-16-36-23/h1,9-12,16,20-21,24,32H,7-8,13-15H2,2-5H3,(H,28,34)(H,30,33)/t20-,21+,24-/m1/s1

Inchi Key

MSSVQZRVBGGJBD-ZFGGDYGUSA-N

Shipping Temperature

Normal

Purity

95%

Storage temp.

Sealed in dry, 2-8C

Country of Origin

CN

Physical Form

Powder or crystals





Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library