Skip to Content
Merck

BL3H9AE4F2D2

BLD Pharmatech

5-((Benzylamino)methyl)-2-((4-ethylbenzyl)amino)-[1,2,4]triazolo[1,5-a]pyrimidin-7-ol

Synonym(s):

5-[(benzylamino)methyl]-2-[(4-ethylbenzyl)amino][1,2,4]triazolo[1,5-a]pyrimidin-7(4H)-one

Select a Size

Change View

Sign in to see your pricing
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Inchi Code

1S/C22H24N6O/c1-2-16-8-10-18(11-9-16)14-24-21-26-22-25-19(12-20(29)28(22)27-21)15-23-13-17-6-4-3-5-7-17/h3-12,23H,2,13-15H2,1H3,(H2,24,25,26,27)

Inchi Key

PAWBAUIXKSTNNX-UHFFFAOYSA-N

Shipping Temperature

Normal

Purity

98%

Country of Origin

CN

Physical Form

Powder or crystals

Storage temp.

Keep in dark place, inert atmosphere, 2-8C



Still not finding the right product?




Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library