Skip to Content
Merck

BL3H9BB32BB3

BLD Pharmatech

3,5-Diamino-N-(N-benzylcarbamimidoyl)-6-chloropyrazine-2-carboxamide

Synonym(s):

N-benzyl-N'-[(3,5-diamino-6-chloro-2-pyrazinyl)carbonyl]guanidine

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Linear Formula:
C13H14O1N7Cl1
CAS Number:
Molecular Weight:
319.75
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Inchi Code

1S/C13H14ClN7O/c14-9-11(16)20-10(15)8(19-9)12(22)21-13(17)18-6-7-4-2-1-3-5-7/h1-5H,6H2,(H4,15,16,20)(H3,17,18,21,22)

Inchi Key

KXDROGADUISDGY-UHFFFAOYSA-N

Shipping Temperature

Normal

Purity

97%

Country of Origin

CN

Physical Form

Powder or crystals

Storage temp.

Keep in dark place, inert atmosphere, 2-8C



Still not finding the right product?




Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library