Skip to Content

COMH93D61DFD

Combi-Blocks, Inc.

2-Hydrazino-5,7-dimethylquinolin-8-ol

Synonym(s):

2-hydrazino-5,7-dimethyl-8-quinolinol

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Linear Formula:
C11H13O1N3
Molecular Weight:
203.24
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Inchi Code

1S/C11H13N3O/c1-6-5-7(2)11(15)10-8(6)3-4-9(13-10)14-12/h3-5,15H,12H2,1-2H3,(H,13,14)

Inchi Key

YMOPKPJDBNRMRC-UHFFFAOYSA-N





Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library