Skip to Content
Merck

EP4966615007

Eppendorf® Bottle Thread Adapter

new

from GL45 to GL32, ETFE

Synonym(s):

thread converter

Sign In to View Organizational & Contract Pricing

Select a Size

Pricing and availability is not currently available.

About This Item

UNSPSC Code:
41121815

Skip To

Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist

material

ETFE

packaging

pack of 1 ea

manufacturer/tradename

Eppendorf® 4966615007

Looking for similar products? Visit Product Comparison Guide

Application

LIQUID HANDLING

Legal Information

Eppendorf is a registered trademark of Eppendorf AG

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service