Skip to Content

System maintenance is scheduled for 11 Sep, 08:00 PM - 12 Sep, 00:00 AM CST [12 Sep 03:00 AM to 07:00 AM CET] and will affect some web and offline transactions, including quotes, price checks, and order status inquiries. We apologize for any inconvenience.

Merck

Skip To

OGS560

PSF-OXB17 - STRONG BACTERIAL PROMOTER VECTOR

plasmid vector for molecular cloning

Synonym(s):

cloning vector, expression vector, molecular cloning vector, plasmid, plasmid vector, snapfast vector, vector

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice

About This Item

NACRES:
NA.85
UNSPSC Code:
12352200
Promoter:
Promoter name: OXB17
Promoter activity: constitutive
Promoter type: bacterial
Origin of replication:
pUC (500 copies)
Bacteria selection:
kanamycin
Reporter gene:
none
Peptide cleavage:
no cleavage
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


form

buffered aqueous solution

mol wt

size 3858 bp

bacteria selection

kanamycin

origin of replication

pUC (500 copies)

peptide cleavage

no cleavage

promoter

Promoter name: OXB17
Promoter activity: constitutive
Promoter type: bacterial

reporter gene

none

shipped in

ambient

storage temp.

−20°C

General description

This is a strong prokaryotic promoter for the production of proteins in E. coli. It was created by modifiying the RecA E. coli promoter to remove the LexA repressor site followed by random mutagensis to reduce expression. In our range of non-inducible promoters this is one of the strongest. Only OXB18 OXB19 and OXB20 are stronger whilst OXB1-OXB16 are weaker.

Promoter Expression Level: This plasmid contains a very strong constitutive E. coli promoter that was derived from the RecA promoter by random mutagenesis. It is part of our constitutive bacterial promoter range. This promoter (OXB17) shows the very high levels of expression in the range with OXB1 showing the lowest level and OXB20 showing the highest level of expression. They require no inducing agent for expression.

Application

Cloning in a gene: This plasmid has been designed to be compatible with a range of cloning techniques. The multiple cloning site contains a range of standard commonly used restriction sites for cloning. Using these sites genes can be inserted using standard cloning methods with DNA ligase. Other methods such as ligase independent cloning (LIC) Gibson Assembly InFusionHD or Seamless GeneArt can also be used and because all of our plasmids are based on the same backbone the same method can be used for cloning into all of our catalogue vectors.

Multiple cloning site notes: There are a few important sites within the MCS. These include the NcoI site the XbaI site and the BsgI and BseRI sites. The NcoI site contains a start codon that is immediately downstream of both a Kozak and Shine-Dalgarno ribosomal binding site. These allow for optimal positioning of genes when the start codon is placed in this location. If this is not required and you wish to use a downstream site for gene cloning you can remove the NcoI site by cleaving the plasmid with KpnI.

The XbaI site contains a stop codon. This stop codon is positioned in a specific position in relation to the BsgI and BseRI sites that are immediately downstream. When either BseRI or BsgI cleave the plasmid they produce a TA overhang from the stop codon in the XbaI site that is compatible with all of our peptide tag plasmids cut with the same sites. BseRI and BsgI sites are non-palindromic and cleave a defined number of bases away from their binding site.

Whenever we clone a gene into our multiple cloning site we always position the start and stop codon in the same positions in the MCS. If the start and ends of the genes are not compatible with NcoI and XbaI we extend the sequence to the nearest external sites but keep the start and stop codons locations consistent.

Analysis Note

To view the Certificate of Analysis for this product, please visit www.oxgene.com

Other Notes

To view sequence information for this product, please visit the product page


Storage Class

12 - Non Combustible Liquids

Flash Point (°F)

Not applicable

Flash Point (°C)

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library




Questions

Reviews

No rating value

Active Filters