MilliporeSigma
  • In vivo pharmacological characterization of TAK-063, a potent and selective phosphodiesterase 10A inhibitor with antipsychotic-like activity in rodents.
In vivo pharmacological characterization of TAK-063, a potent and selective phosphodiesterase 10A inhibitor with antipsychotic-like activity in rodents.
The Journal of pharmacology and experimental therapeutics (2014-12-20)
Kazunori Suzuki, Akina Harada, Eri Shiraishi, Haruhide Kimura
ABSTRACT
Phosphodiesterase 10A (PDE10A) is a cAMP/cGMP phosphodiesterase highly expressed in medium spiny neurons (MSNs) in the striatum. We evaluated the in vivo pharmacological profile of a potent and selective PDE10A inhibitor, TAK-063 (1-[2-fluoro-4-(1H-pyrazol-1-yl)phenyl]-5-methoxy-3-(1-phenyl-1H-pyrazol-5-yl)-pyridazin-4(1H)-one). TAK-063 at 0.3 and 1 mg/kg p.o., increased cAMP and cGMP levels in the rodent striatum and upregulated phosphorylation levels of key substrates of cAMP- and cGMP-dependent protein kinases. TAK-063 at 0.3 and 1 mg/kg p.o., strongly suppressed MK-801 [(5R,10S)-(+)-5-methyl-10,11-dihydro-5H-dibenzo[a,d]cyclohepten-5,10-imine]-induced hyperlocomotion, which is often used as a predictive model for antipsychotic-like activity in rodents. Upregulation of striatal cAMP/cGMP levels and the antipsychotic-like effect of TAK-063 were not attenuated after 15 days of pretreatment with TAK-063 in mice. The potential side effect profile of TAK-063 was assessed in rats using the clinical antipsychotics haloperidol, olanzapine, and aripiprazole as controls. TAK-063 did not affect plasma prolactin or glucose levels at doses up to 3 mg/kg p.o. At 3 mg/kg p.o., TAK-063 elicited a weak cataleptic response compared with haloperidol and olanzapine. Evaluation of pathway-specific markers (substance P mRNA for the direct pathway and enkephalin mRNA for the indirect pathway) revealed that TAK-063 activated both the direct and indirect pathways of MSNs. These findings suggest that TAK-063 represents a promising drug for the treatment of schizophrenia with potential for superior safety and tolerability profiles.
Product Description
Haloperidol, European Pharmacopoeia (EP) Reference Standard
Pricing and availability is not currently available.
Sigma-Aldrich
Bicinchoninic acid disodium salt hydrate, Vetec, reagent grade, 98%
Pricing and availability is not currently available.
USP
Haloperidol, United States Pharmacopeia (USP) Reference Standard
Pricing and availability is not currently available.
Sigma-Aldrich
Bicinchoninic acid disodium salt hydrate, ≥98% (HPLC)
Pricing and availability is not currently available.
Sigma-Aldrich
Haloperidol, powder
Pricing and availability is not currently available.
Aripiprazole, European Pharmacopoeia (EP) Reference Standard
Pricing and availability is not currently available.
Sigma-Aldrich
Aripiprazole, ≥98% (HPLC)
Pricing and availability is not currently available.
Haloperidol for peak identification, European Pharmacopoeia (EP) Reference Standard
Pricing and availability is not currently available.
Haloperidol for system suitability, European Pharmacopoeia (EP) Reference Standard
Pricing and availability is not currently available.
Sigma-Aldrich
Olanzapine, ≥98% (HPLC)
Pricing and availability is not currently available.
Sigma-Aldrich
Hydrogen bromide, ReagentPlus®, ≥99%
Pricing and availability is not currently available.
Sigma-Aldrich
Rolipram, solid, ≥98% (HPLC)
Pricing and availability is not currently available.
Sigma-Aldrich
Hydrobromic acid, 48 wt. % in H2O, ≥99.99%
Pricing and availability is not currently available.
Sigma-Aldrich
Hydrobromic acid solution, 33 wt. % in acetic acid
Pricing and availability is not currently available.
Sigma-Aldrich
Hydrogen bromide solution, 33 wt. % in acetic acid
Pricing and availability is not currently available.
Sigma-Aldrich
Hydrobromic acid, reagent grade, 48%
Pricing and availability is not currently available.
Sigma-Aldrich
Hydrobromic acid, ACS reagent, 48%
Pricing and availability is not currently available.
Olanzapine, European Pharmacopoeia (EP) Reference Standard
Pricing and availability is not currently available.
Olanzapine for system suitability, European Pharmacopoeia (EP) Reference Standard
Pricing and availability is not currently available.
USP
Olanzapine, United States Pharmacopeia (USP) Reference Standard
Pricing and availability is not currently available.