Skip to Content
Merck

ATE514316594

AstaTech, Inc.

METHYL 5-METHYL-2-HEXENOATE

Sign In to View Organizational & Contract Pricing

Select a Size


About This Item

CAS Number:
Molecular Weight:
142.2
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist

bp

95-100 C/26 mmHg

Iupac Name

methyl (2E)-5-methyl-2-hexenoate

Inchi Code

1S/C8H14O2/c1-7(2)5-4-6-8(9)10-3/h4,6-7H,5H2,1-3H3/b6-4+

InChI key

KNSMNULJDOTFMN-GQCTYLIASA-N

storage temp.

2-8 C

Purity

95%

Country of Origin

CN

Shipping Temperature

Normal

Physical Form

Liquid

Product Link

https://astatechinc.com/CPSResult.php?CRNO=80879

Flash Point

57 C/134 F

Boiling point

95-100 C/26 mmHg

Signal Word

Warning

Hazard Statements


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service