Skip to Content
Merck

BDPH9BCC2D85

Bide Pharmatech

4-Bromo-2-(trifluoromethyl)phenol

Synonym(s):

4-bromo-2-(trifluoromethyl)phenol

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Linear Formula:
C7H4O1Br1F3
Molecular Weight:
241.01
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Inchi Code

1S/C7H4BrF3O/c8-4-1-2-6(12)5(3-4)7(9,10)11/h1-3,12H

Inchi Key

PDPGERGWEOJVDC-UHFFFAOYSA-N





Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library