Skip to Content
Merck

BDPH9BCCE91B

Bide Pharmatech

Ethyl 2-(6-methylpyridin-2-yl)acetate

Synonym(s):

ethyl (6-methyl-2-pyridinyl)acetate

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Linear Formula:
C10H13O2N1
CAS Number:
Molecular Weight:
179.22
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Inchi Code

1S/C10H13NO2/c1-3-13-10(12)7-9-6-4-5-8(2)11-9/h4-6H,3,7H2,1-2H3

Inchi Key

AJXKDBXXGPKKPN-UHFFFAOYSA-N

Shipping Temperature

Normal

Purity

95%

Storage temp.

Inert atmosphere, room temperature

Country of Origin

CN

Physical Form

Colorless to Yellow-brown to Red Liquid



Still not finding the right product?


Precautionary Statements

Signal Word

Warning

Pictograms

Exclamation mark


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library