Sign In to View Organizational & Contract Pricing.
Select a Size
Change View
| Size/SKU | Availability | Price |
|---|
About This Item
Pricing and availability is not currently available. Contact Technical Service
Inchi Code
1S/C5H13N3.2ClH/c6-8-5-1-3-7-4-2-5;;/h5,7-8H,1-4,6H2;2*1H
Inchi Key
AFLRARNCCZQUGM-UHFFFAOYSA-N
Shipping Temperature
Normal
Purity
95%
Storage temp.
Sealed in dry, 2-8C
Country of Origin
CN
Physical Form
White to Yellow Solid
Still not finding the right product?
Explore all of our products under 4-Hydrazinylpiperidine dihydrochloride
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
