Skip to Content
Merck

BDPH9BCCFF06

Bide Pharmatech

Methyl 3-aminobenzo[b]thiophene-2-carboxylate

Synonym(s):

methyl 3-amino-1-benzothiophene-2-carboxylate

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Linear Formula:
C10H9O2N1S1
CAS Number:
Molecular Weight:
207.25
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Inchi Code

1S/C10H9NO2S/c1-13-10(12)9-8(11)6-4-2-3-5-7(6)14-9/h2-5H,11H2,1H3

Inchi Key

VLHHEYMZLXKSQO-UHFFFAOYSA-N

Shipping Temperature

Normal

Purity

97%

Storage temp.

Keep in dark place, inert atmosphere, room temperature

Country of Origin

CN

Physical Form

Light-yellow to yellow-brown to dark brown powder or crystals



Still not finding the right product?


Signal Word

Warning

Pictograms

Exclamation mark


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library