Skip to Content

BDPH9C021E55

Bide Pharmatech

N-(2-Fluorophenyl)cinnamamide

Synonym(s):

(2E)-N-(2-fluorophenyl)-3-phenyl-2-propenamide

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Linear Formula:
C15H12O1N1F1
CAS Number:
Molecular Weight:
241.26
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Inchi Code

1S/C15H12FNO/c16-13-8-4-5-9-14(13)17-15(18)11-10-12-6-2-1-3-7-12/h1-11H,(H,17,18)/b11-10+

Inchi Key

GFAVMCPSQILRPK-ZHACJKMWSA-N

Shipping Temperature

Normal

Purity

98%

Storage temp.

Sealed in dry, room temperature

Country of Origin

CN

Physical Form

Solid



Still not finding the right product?

Explore all of our products under N-(2-Fluorophenyl)cinnamamide



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library