Sign In to View Organizational & Contract Pricing.
Select a Size
Change View
| Size/SKU | Availability | Price |
|---|
About This Item
NACRES:
NB.79
UNSPSC Code:
41116129
Pricing and availability is not currently available.
manufacturer/tradename
Milliflex®
input
sample type pharmaceutical(s)
sample type beer
sample type cosmetics
sample type bottled water
application(s)
bioburden testing
cosmetics
pharmaceutical
water monitoring
shipped in
ambient
General description
The compact Milliflex Plus vacuum pump streamlines filtration to accelerate QC testing. Together, the Milliflex filter units and the Milliflex Plus vacuum pump provide an integrated solution to deliver reliable and accurate results in high throughput production environments.
Sanitization is easy, so there’s minimal disruption to workflow. For even greater throughput, a high-volume workstation of up to three Milliflex PLUS pumps can be set-up and used by a single operator. Testing reveals 3 Milliflex PLUS pumps are faster than a 6-place manifold.
Support and Services
{HCompany} provides technical support and a comprehensive services offering to help you save your QC resources and facilitate the implementation of your MilliFlex Plus Pump Single unit into your daily testing routine. Our highly experienced teams are there to assist you.
Sanitization is easy, so there’s minimal disruption to workflow. For even greater throughput, a high-volume workstation of up to three Milliflex PLUS pumps can be set-up and used by a single operator. Testing reveals 3 Milliflex PLUS pumps are faster than a 6-place manifold.
Support and Services
{HCompany} provides technical support and a comprehensive services offering to help you save your QC resources and facilitate the implementation of your MilliFlex Plus Pump Single unit into your daily testing routine. Our highly experienced teams are there to assist you.
Application
Beer, Bottled Water, Cosmetics, Pharmaceutical Products Analysis, Bioburden Testing, Water Monitoring
Legal Information
MILLIFLEX is a registered trademark of Merck KGaA, Darmstadt, Germany
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.