Skip to Content
Merck

BPS78366

PARP Substrate Mixture 1

Synonym(s):

PARP Substrate Mixture

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

UNSPSC Code:
47101613
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


manufacturer/tradename

BPS Bioscience

storage temp.

-65 to -96°C

General description

Proprietary mixture including biotinylated and non-biotinylated NAD+, optimized for use with BPS Bioscience′s poly(ADP) ribose polymerase (PARP) assay kits:
  • PARP1 Chemiluminescent Assay Kit (BPS Bioscience #80551)
  • PARP2 Chemiluminescent Assay Kit (BPS Bioscience #80552)
  • PARP3 Chemiluminescent Assay Kit (BPS Bioscience #80553)
  • PARP10 Chemiluminescent Assay Kit (BPS Bioscience #80560)

Application

Suitable for use with cell extracts and purified PARP enzymes.

Datasheet and Protocols may be found here.

Preparation Note

Tris-buffered solution

Analysis Note

Avoid freeze/thaw cycles.


Storage Class

10 - Combustible liquids

Flash Point (°F)

Not applicable

Flash Point (°C)

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library