- Nylon membrane as a fluorimetric probe for the herbicide bentazone.
Nylon membrane as a fluorimetric probe for the herbicide bentazone.
Analytical and bioanalytical chemistry (2012-01-10)
Jessica P Chiarandini, Graciela M Escandar
PMID22227748
ABSTRACT
The fluorimetric signal produced by bentazone retained in selected solid surfaces was investigated. Among the different tested supports, only a microporous nylon membrane produced the desired signal. The quantitative study was carried out by second-order calibration using parallel factor analysis, allowing the determination in a highly interfering medium. A detection limit of 0.4 ng mL(-1), a prediction relative error of 8%, and a sample frequency of ten samples per hour were obtained in spiked natural waters using green analytical chemistry principles.