- [Effects of propham and chlorpropham on the embryonic development of various vertebrates].
[Effects of propham and chlorpropham on the embryonic development of various vertebrates].
Comptes rendus des seances de la Societe de biologie et de ses filiales (1981-01-01)
B Caporiccio, R Michel, J Tournamille, P Sentein
PMID6459825
ABSTRACT
The effect of propham and chlorpropham on Amphibians embryonic development changes with doses as well as species. The action of C.I.P.C. is always determinative. The egg poisoned in albumen involves important chick embryo malformations, but the development is stopped by the yolky vesicle poisoning. The intraperitoneal injection of C.I.P.C., strongly disturbs fetus physiology in Mouse.